ENAH38A2CC7B Enamine

1-bromo-4,4-diethoxy-1,1,2,2-tetrafluorobutane

  • MDL number MFCD31810880
  • Cas Number : 2305255-43-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6758877

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6758877
Availability details for
Quantity
  • Availability
  • Add to Cart