ENAH38A2CC9D Enamine

3-(dimethylamino)-2,2-difluoropropan-1-ol hydrochloride

  • MDL number MFCD31811151
  • Cas Number : 2305252-90-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6759676

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6759676
Availability details for
Quantity
  • Availability
  • Add to Cart