ENAH38A2CCA5 Enamine

3-(iodomethyl)-2,8-dioxaspiro[4.5]decan-1-one

  • MDL number MFCD31811010
  • Cas Number : 2246354-70-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6759855

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6759855
Availability details for
Quantity
  • Availability
  • Add to Cart