ENAH38A2CCA8 Enamine

2-(1-{[(9H-fluoren-9-yl)methoxy]carbonyl}-3,3-dimethylpiperidin-4-yl)acetic acid

  • MDL number MFCD31811049
  • Cas Number : 2305255-17-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6759871

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6759871
Availability details for
Quantity
  • Availability
  • Add to Cart