ENAH38A2CCAB Enamine

4-[4-(4-chlorobenzoyl)phenoxy]butanoic acid

  • MDL number MFCD31811056
  • Cas Number : 2305255-96-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6760082

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6760082
Availability details for
Quantity
  • Availability
  • Add to Cart