ENAH38A2CCAE Enamine

ethyl (1R,3r,5S)-6-oxabicyclo[3.1.0]hexane-3-carboxylate

  • MDL number MFCD31811164
  • Cas Number : 1622218-77-2
  • Molecular Weight : 156.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6760207

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6760207
Availability details for
Quantity
  • Availability
  • Add to Cart