ENAH38A2CCB3 Enamine

tert-butyl N-[1-(2-methanesulfonyl-6-phenoxypyrimidin-4-yl)piperidin-4-yl]carbamate

  • MDL number MFCD31811114
  • Cas Number : 2305255-24-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6760300

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6760300
Availability details for
Quantity
  • Availability
  • Add to Cart