ENAH38A2CCDC Enamine

1-(phenoxymethyl)-4-(propan-2-yl)benzene

  • MDL number MFCD21366680
  • Cas Number : 1457318-82-9
  • Molecular Weight : 226.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-6735627

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6735627
Availability details for
Quantity
  • Availability
  • Add to Cart