ENAH38A2CCED Enamine

tert-butyl 4-methoxy-3,10-diazabicyclo[4.3.1]dec-3-ene-10-carboxylate

  • MDL number MFCD31810902
  • Cas Number : 2305256-05-5
  • Molecular Weight : 268.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-6736280

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6736280
Availability details for
Quantity
  • Availability
  • Add to Cart