ENAH38A2CD2B Enamine

5-amino-3-(propan-2-yl)-1H-pyrazole-4-carboxamide

  • MDL number MFCD31810684

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6739206

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6739206
Availability details for
Quantity
  • Availability
  • Add to Cart