ENAH38A2CD3A Enamine

N,N-bis(prop-2-en-1-yl)carbamoyl chloride

  • MDL number MFCD02093718
  • Cas Number : 25761-72-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6739665

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6739665
Availability details for
Quantity
  • Availability
  • Add to Cart