ENAH38A2CD3B Enamine

tert-butyl (2R,6S)-4-(chlorosulfonyl)-2,6-dimethylpiperazine-1-carboxylate

  • MDL number MFCD31915119
  • Cas Number : 2309432-95-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6739734

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6739734
Availability details for
Quantity
  • Availability
  • Add to Cart