ENAH38A2CD3D Enamine

tert-butyl 2,6-dioxo-9-azabicyclo[3.3.1]nonane-9-carboxylate

  • MDL number MFCD31810946
  • Cas Number : 1434905-61-9
  • Molecular Weight : 253.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-6739818

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6739818
Availability details for
Quantity
  • Availability
  • Add to Cart