ENAH38A2CD47 Enamine

2-(dimethylphosphoryl)pyridin-4-amine

  • MDL number MFCD31915712
  • Cas Number : 2309447-77-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6740378

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6740378
Availability details for
Quantity
  • Availability
  • Add to Cart