ENAH38A2CD50 Enamine

1-(bromodifluoromethyl)-2-fluorobenzene

  • MDL number MFCD31810953
  • Cas Number : 2305255-13-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6740623

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6740623
Availability details for
Quantity
  • Availability
  • Add to Cart