ENAH38A2CD51 Enamine

1-(methoxymethyl)-2-(tributylstannyl)-1H-imidazole

  • MDL number MFCD31810954

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6740639

Safety Information

signalword Danger
pcodes P260,P264,P270,P273,P280,P301+P310,P302+P352,P305+P351+P338,P312,P314,P330,P332+P313,P337+P313,P362,P363,P391,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-6740639
pictograms GHS06,GHS08,GHS09
hstatements H301,H312,H315,H319,H372,H410
Availability details for
Quantity
  • Availability
  • Add to Cart