ENAH38A2CD5D Enamine

3,3,4,4-tetrafluorobutyl methanesulfonate

  • MDL number MFCD31915165
  • Cas Number : 2309455-55-6
  • Molecular Weight : 224.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6740940

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6740940
Availability details for
Quantity
  • Availability
  • Add to Cart