ENAH38A2CD5F Enamine

3-(1-{[(tert-butoxy)carbonyl]amino}cyclopentyl)prop-2-ynoic acid

  • MDL number MFCD31810956
  • Cas Number : 2305255-33-6
  • Molecular Weight : 253.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6742975

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6742975
Availability details for
Quantity
  • Availability
  • Add to Cart