ENAH38A2CD69 Enamine

1,3,7-trichloroisoquinoline

  • MDL number MFCD11112091
  • Cas Number : 21902-41-0
  • Molecular Weight : 232.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6743373

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6743373
Availability details for
Quantity
  • Availability
  • Add to Cart