ENAH38A2CD74 Enamine

6-methoxy-1-(naphthalen-1-yl)-1H,2H,3H,4H,9H-pyrido[3,4-b]indole

  • MDL number MFCD05190807
  • Cas Number : 529476-44-6
  • Molecular Weight : 328.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6743746

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6743746
Availability details for
Quantity
  • Availability
  • Add to Cart