ENAH38A2CD79 Enamine

methyl 5-(cyclopropylmethyl)-1,2-oxazole-3-carboxylate

  • MDL number MFCD31915716
  • Cas Number : 2309456-83-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6744025

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6744025
Availability details for
Quantity
  • Availability
  • Add to Cart