ENAH38A2CD7D Enamine

5-aminopyrimidine-4,6-dithiol

  • MDL number MFCD19301129
  • Cas Number : 54660-92-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6744121

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6744121
Availability details for
Quantity
  • Availability
  • Add to Cart