ENAH38A2CD87 Enamine

2-{spiro[3.3]heptan-1-yl}acetic acid

  • MDL number MFCD31915172
  • Cas Number : 2309457-81-4
  • Molecular Weight : 154.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6745280

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6745280
Availability details for
Quantity
  • Availability
  • Add to Cart