ENAH38A2CD90 Enamine

(4S)-4-{[(tert-butoxy)carbonyl]amino}pentanoic acid

  • MDL number MFCD06410963
  • Cas Number : 207924-92-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6745652

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6745652
Availability details for
Quantity
  • Availability
  • Add to Cart