ENAH38A2CD9A Enamine

1-bromo-5-(bromomethyl)-3-chloro-2-methylbenzene

  • MDL number MFCD31915177
  • Cas Number : 2091468-76-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6745859

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6745859
Availability details for
Quantity
  • Availability
  • Add to Cart