ENAH38A2CDA1 Enamine

2-(aminomethyl)-6,6-difluorospiro[3.3]heptane-2-carboxylic acid

  • MDL number MFCD31915181
  • Cas Number : 2309460-18-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6746096

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6746096
Availability details for
Quantity
  • Availability
  • Add to Cart