ENAH38A2CDA2 Enamine

tert-butyl 4-acetamido-4-methylpiperidine-1-carboxylate

  • MDL number MFCD19707618
  • Cas Number : 1673527-45-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form oil
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6746153

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6746153
Availability details for
Quantity
  • Availability
  • Add to Cart