ENAH38A2CDA3 Enamine

[(R)-amino(phenyl)methyl]phosphonic acid hydrochloride

  • MDL number MFCD31810705
  • Cas Number : 411231-12-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6746172

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6746172
Availability details for
Quantity
  • Availability
  • Add to Cart