ENAH38A2CDAE Enamine

[amino(diphenylamino)methylidene]azanium iodide

  • Cas Number : 2361775-33-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data I-
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6746436

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6746436
Availability details for
Quantity
  • Availability
  • Add to Cart