ENAH38A2CDD1 Enamine

1-(1H-pyrazol-4-yl)-1,3-diazinane-2,4-dione

  • MDL number MFCD31810966
  • Cas Number : 2305255-85-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6748204

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6748204
Availability details for
Quantity
  • Availability
  • Add to Cart