ENAH38A2CDE3 Enamine

7-chloro-8-methoxy-1,2,3,4-tetrahydroquinoline

  • MDL number MFCD31810969
  • Cas Number : 1782455-38-2
  • Molecular Weight : 197.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6748390

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6748390
Availability details for
Quantity
  • Availability
  • Add to Cart