ENAH38A2CDEB Enamine

2-[1-(4-fluorophenyl)-2,5-dimethyl-1H-pyrrol-3-yl]acetic acid

  • MDL number MFCD01688622
  • Cas Number : 42779-83-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6748478

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6748478
Availability details for
Quantity
  • Availability
  • Add to Cart