ENAH38A2CDF0 Enamine

1-(dimethylphosphoryl)-4-fluorobenzene

  • MDL number MFCD31915098
  • Cas Number : 18437-74-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6748502

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6748502
Availability details for
Quantity
  • Availability
  • Add to Cart