ENAH38A2CDF1 Enamine

tert-butyl 2-(2,5-dioxopyrrolidin-3-yl)acetate

  • MDL number MFCD31810730
  • Cas Number : 2305255-32-5
  • Molecular Weight : 213.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6748525

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6748525
Availability details for
Quantity
  • Availability
  • Add to Cart