ENAH38A2CDF4 Enamine

4-(dimethylphosphoryl)-1-fluoro-2-nitrobenzene

  • MDL number MFCD31810536
  • Cas Number : 2287314-01-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6748549

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6748549
Availability details for
Quantity
  • Availability
  • Add to Cart