ENAH38A2CDF6 Enamine

N'-hydroxy-2-(2-oxopyrrolidin-1-yl)ethanimidamide

  • Cas Number : 126145-44-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6748568

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6748568
Availability details for
Quantity
  • Availability
  • Add to Cart