ENAH38A2CDFD Enamine

tert-butyl 3-isothiocyanatopyrrolidine-1-carboxylate

  • MDL number MFCD31810734
  • Cas Number : 1081979-18-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form oil
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6748633

Safety Information

signalword Danger
pcodes P260,P261,P264,P270,P271,P280,P301+P310,P301+P330+P331,P302+P352,P303+P361+P353,P304+P340,P305+P351+P338,P310,P311,P312,P330,P361,P363,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-6748633
pictograms GHS05,GHS06
hstatements H301,H311,H314,H331
Availability details for
Quantity
  • Availability
  • Add to Cart