ENAH38A2CE05 Enamine

3-(6-bromopyridin-3-yl)-2-{[(tert-butoxy)carbonyl]amino}propanoic acid

  • MDL number MFCD09916885
  • Cas Number : 1379891-42-5
  • Molecular Weight : 345.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-6504597

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6504597
Availability details for
Quantity
  • Availability
  • Add to Cart