ENAH38A2CE09 Enamine

4-(2-amino-6-methyl-4-oxo-1,4,5,6-tetrahydropyridin-3-yl)-1-methylpyridin-1-ium hydrochloride chloride

  • MDL number MFCD31915774

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data Cl.[Cl-]
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6748682

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6748682
Availability details for
Quantity
  • Availability
  • Add to Cart