ENAH38A2CE15 Enamine

{1-[(dimethylamino)methyl]cyclobutyl}methanamine dihydrochloride

  • MDL number MFCD28894738
  • Cas Number : 2305252-52-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6748764

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6748764
Availability details for
Quantity
  • Availability
  • Add to Cart