ENAH38A2CE1D Enamine

1-iodo-2-(prop-2-yn-1-yloxy)benzene

  • MDL number MFCD00973039
  • Cas Number : 41876-99-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 40-41
Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6748797

Safety Information

signalword Danger
pcodes P261,P264,P270,P271,P272,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P310,P312,P330,P332+P313,P333+P313,P362,P363,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-6748797
pictograms GHS05,GHS07
hstatements H302,H312,H315,H317,H318,H332,H335
Availability details for
Quantity
  • Availability
  • Add to Cart