ENAH38A2CE29 Enamine

1-methyl-5-(trifluoromethyl)-1H,4H,5H,6H,7H-pyrazolo[4,3-b]pyridine-6-carboxylic acid

  • MDL number MFCD31810758
  • Cas Number : 2305253-10-3
  • Molecular Weight : 249.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6748912

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6748912
Availability details for
Quantity
  • Availability
  • Add to Cart