ENAH38A2CE2D Enamine

2-(3-amino-1-oxo-1,5,6,7,8,8a-hexahydroindolizin-2-yl)-1-methylpyridin-1-ium chloride

  • MDL number MFCD31915776

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data Cl-
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6748949

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6748949
Availability details for
Quantity
  • Availability
  • Add to Cart