ENAH38A2CE39 Enamine

(3R,4S)-3,4-dihydroxy-1lambda6-thiolane-1,1-dione

  • MDL number MFCD00154859
  • Cas Number : 36802-08-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6749037

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6749037
Availability details for
Quantity
  • Availability
  • Add to Cart