ENAH38A2CE43 Enamine

6-bromo-3-iodo-2-methylpyrazolo[1,5-a]pyrimidine hydrochloride

  • MDL number MFCD31811031
  • Cas Number : 2305254-62-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6750813

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6750813
Availability details for
Quantity
  • Availability
  • Add to Cart