ENAH38A2CE45 Enamine

2-amino-1-(1,3-dimethyl-1H-pyrazol-4-yl)ethan-1-one hydrochloride

  • MDL number MFCD31810765
  • Cas Number : 2305254-95-7
  • Molecular Weight : 189.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6750816

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6750816
Availability details for
Quantity
  • Availability
  • Add to Cart