ENAH38A2CE55 Enamine

methyl 4-methyl-6-oxopiperidine-3-carboxylate

  • MDL number MFCD31811036
  • Cas Number : 1782536-27-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6750982

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6750982
Availability details for
Quantity
  • Availability
  • Add to Cart