ENAH38A2CE57 Enamine

(1r,3r)-3-(2-methoxyphenyl)cyclobutan-1-amine hydrochloride

  • MDL number MFCD31810775
  • Cas Number : 2305151-95-3
  • Molecular Weight : 213.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6750999

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6750999
Availability details for
Quantity
  • Availability
  • Add to Cart