ENAH38A2CE58 Enamine

tert-butyl N-{[(1r,3r)-3-(aminomethyl)-1-fluorocyclobutyl]methyl}carbamate

  • MDL number MFCD31915341
  • Cas Number : 2309468-27-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form oil
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6751003

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6751003
Availability details for
Quantity
  • Availability
  • Add to Cart