ENAH38A2CE71 Enamine

(3R)-3,6-bis({[(tert-butoxy)carbonyl]amino})hexanoic acid

  • MDL number MFCD31915488
  • Cas Number : 102686-48-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-6754326

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-6754326
Availability details for
Quantity
  • Availability
  • Add to Cart